AdvanTech TRIonic™ 514 Tripolymer Controlled Environment Gloves
Actual product may vary
AdvanTech TRIonic™ 514 Tripolymer Controlled Environment Gloves
AdvanTech TRIonic™ 514 Tripolymer Controlled Environment Gloves
MAPA
AdvanTech TRIonic™ 514 Tripolymer Controlled Environment Gloves
Extra long 15 in. tripolymer cleanroom gloves provide optimal chemical resistance and are recommended for Class 10/ ISO Class 4 critical environments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-249-5377
FIS19-249-5366
Catalog number FIS19-249-537
Price (SGD)
863.00
Case
Size:
7
Price (SGD)
863.00
Case
Tripolymer blend (nitrile, neoprene and natural rubber) outperforms natural rubber in applications involving aggressive chemicals
  • Natural rubber protein levels test below detection
  • Strong and durable; highly resistant to tears, snags, cuts
  • Unpigmented with diamond grip
  • Unlined and 20 mil thick, flat-cuffed
  • Tapered cuff helps keep glove up over forearm
Specifications
Certifications/ComplianceFDA
Certifications/Compliance (Europe)EN388, EN ISO 374-1, EN ISO 374-5
Cleanroom ClassClass 10 (ISO 4)
ColorNatural
Exterior FinishFully Textured
HandSpecific
Latex FreeNo
MaterialTripolymer
Product TypeControlled Environment Gloves
Size7
SterilityNon-sterile
Thickness (English)20 mil
Thickness (Metric)0.50 mm
Model514317
Packaging Type1 pair/bag; 72 pairs/carton
SeriesAdvanTech™ 514
Length (English)15 in.
Length (Metric)38 cm
Unit SizeCase