Chemical Splash and Impact Goggles
Actual product may vary
Chemical Splash and Impact Goggles
Lens coated to minimize scratching and fogging.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeVentilation DetailsCertifications/Compliance
FIS19-181-504Chemical Splash Protection GogglesFour indirect ventsANSI/ISEA Z87.1-2020: Z87+ U6 D3
FIS19-181-503Impact GogglesPerforations on goggle top and sides provide direct ventilationANSI/ISEA Z87.1-2020: Z87+ U6
Catalog number FIS19-181-504
Price (SGD)
6.00
Each
Product Type:
Chemical Splash Protection Goggles
Ventilation Details:
Four indirect vents
Certifications/Compliance:
ANSI/ISEA Z87.1-2020: Z87+ U6 D3
Price (SGD)
6.00
Each
Lens coated to minimize scratching and fogging
  • Built for comfortable protection with a soft, flexible PVC body that conforms to facial contours
  • Large frames fit over most prescription eyewear
  • Coated, clear polycarbonate lens resists impact, scratching and fogging
  • Headstrap easily adjusts for a custom fit
  • Protect eyes from dust, mist and other airborne particles
  • Perforations on goggle top and sides provide direct ventilation
Specifications
ShapeRound
Lens CoatingAnti-fog, Scratch-resistant
Certifications/ComplianceANSI/ISEA Z87.1-2020: Z87+ U6 D3
Lens TintClear
Product TypeChemical Splash Protection Goggles
Frame ColorGreen
Ventilation DetailsFour indirect vents
MaterialPolycarbonate
Lens MaterialPolycarbonate
Frame MaterialPVC
Unit SizeEach