▸
Personal Protective Equipment
▸
Safety Clothing
▸
Protective Sleeves
▸
Protective Polyethylene Sleeve Covers
Actual product may vary
Fisherbrand
Protective Polyethylene Sleeve Covers
Disposable general purpose sleeves
Have Questions?
Catalog number
FIS19-170-904
Price (SGD)
-
Disposable general purpose sleeves
Polyethylene
Length: 18 in. (45.7cm)
Latex-free
Blue
Thickness: 6 mil (0.15mm)
Elastic cuffs
Specifications
Product Type
Sleeve Cover
Material
Polyethylene
Color
Blue
Cuff Style
Elastic
Latex Free
Yes
Length (English)
18 in.
Length (Metric)
45.7 cm