Wooden Cuticle/Orange Stick
Wooden Cuticle/Orange Stick
Actual product may vary
Puritan
Wooden Cuticle/Orange Stick
Wooden cuticle or orange stick with single bevel and double bevel on opposite ends.
Have Questions?
Catalog NumberLength (Metric)
FIS19-166-77217.7 cm
Catalog number FIS19-166-772
Price (SGD)
-
Length (Metric):
17.7 cm
Used in assembly and repairing of circuit board, general electronics manufacturing for probing loose wires and positioning small parts; Used in podiatry, health and beauty applications; Dimensions: 7 in. x 0.156 in.; Made in USA
Specifications
Certifications/ComplianceISO 9001:2008, ISO 13485:2003, FDA GMP's
Length (English)7 in.
For Use With (Application)Assembly and repair of circuit boards, general electronics, probing loose wires, positioning small parts
Product TypeStick
Diameter (Metric) Handle0.39 cm
Length (Metric)17.7 cm
Diameter (English) Tip0.156 in.