Men's Staff Coats
Actual product may vary
Men's Staff Coats
Men's Staff Coats
Men's Staff Coats
VF
Men's Staff Coats
Men's long sleeve staff coat features a pleated-back style for non-restricted movement, vents to increase mobility, and a sewn-on belt for an adjustable fit.
Have Questions?
Catalog NumberSizeStyleSize (English) Chest
FIS19-166-2552X-LargeRegular52 to 54 in.
Catalog number FIS19-166-255
Price (SGD)
106.00
Each
Size:
2X-Large
Style:
Regular
Size (English) Chest:
52 to 54 in.
Price (SGD)
106.00
Each
  • Primary Closure: Button
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: 6 oz. Fine-line Twill
  • Blend: 80% Polyester/20% Combed cotton
  • Finish: Wrinkle Resistant
  • Closure: Four non-yellowing UV buttons
  • Collar: One-piece, notched lapel
  • Length: 39 in. (based on size medium - back)
  • Pocket: Left exterior chest pocket, Bartacked pencil stall, Two lower exterior pockets
  • Features: Non-yellowing buttons for long-lasting whiteness and professional appearance
  • Requires minimum ironing thanks to a wrinkle-resistant finish
  • One chest pocket and two lower pockets provide storage for tools and personal items
  • Side vent openings for easy access to underneath attire
  • Pleated back with sewn-on belt for a fitted, professional appearance
  • Country of Origin: Imported, Imported Fabric
  • Fit Details: Fits true to size. Meant to be worn over clothes.
  • Gender: Male
  • Materials: Twill, Cotton/Polyester
Specifications
Product TypeGeneral Purpose Lab Coat
Size2X-Large
GenderMen's
MaterialCotton, Polyester
ColorWhite
Closure TypeButtons
DisposableReusable
SterilityNon-sterile
ResistanceWrinkles
StyleRegular
No. of Pockets3
Collar TypeNotched Lapel
FinishPre-cure Durable Press
Size (English) Chest52 to 54 in.
Size (Metric) Chest132 to 137 cm
Length (English)39 in.
Length (Metric)99.06 cm
ConstructionPolyester, Combed Cotton
Unit SizeEach