MICROFLEX™ HSCE4-879 Sterile Latex Gloves
Actual product may vary
Microflex
MICROFLEX™ HSCE4-879 Sterile Latex Gloves
Powder free latex gloves are hand-specific ro reduce stress and strain. Recommended for Class 10 /ISO 4 cleanrooms.
Have Questions?
Catalog NumberSize
FIS19-166-12558
Catalog number FIS19-166-1255
Price (SGD)
1,550.00
Case
Size:
8
Price (SGD)
1,550.00
Case
Class 10 Hand-Specific Sterile Latex Gloves
Produced in totally integrated facility certified to ISO 9001 standards
  • Micro-textured surface enhances wet or dry grip
  • Does not contain silcone
  • Irradiation indicators placed on all master poly bags and cartons
  • Certified sterile at a minimum dosage of 25Kgy
  • Hand-specific
  • Thickness: 7.9 mil at fingertip
  • Length: 11 in. (28 cm)
  • Natural color
Specifications
Certifications/ComplianceAAMI, ASTM D6319-10, IEST-RP-CC005.3, FTMS 101-C, 4046
Certifications/Compliance (Europe)EN 455 1-4
Cleanroom ClassClass 10 (ISO 4)
AQL1.5
ColorNatural
DisposableSingle-use
Exterior FinishNo Texture
Finger Thickness (English)7.9 mil
Finger Thickness (Metric)0.20 mm
HandSpecific
MaterialLatex
Palm Thickness (English)5.9 mil
Palm Thickness (Metric)0.15 mm
PowderedPowder Free
Size8
SterilitySterile
Thickness (English)7.9 mil
Thickness (Metric)0.20 mm
Manufacturing/QMS Audit StandardsISO2004, ISO9001
ModelHSCE4-879-80
SeriesMICROFLEX™ HSCE4-879
Length (English)12 in.
Length (Metric)30.5 cm
Unit SizeCase