Neoprene Quick-Dry Foam Squeegees
Actual product may vary
Perfex
Neoprene Quick-Dry Foam Squeegees
Flexible blades are specifically designed for use on uneven or irregular surfaces
Have Questions?
Catalog NumberMaterial
FIS19-165-126Neoprene Rubber, Polymer Channel
Catalog number FIS19-165-126
Price (SGD)
1,038.00
Pack
Material:
Neoprene Rubber, Polymer Channel
Price (SGD)
1,038.00
Pack
Twin-blade floor squeegee made of neoprene rubber with a durable polymer channel
  • L-N-T connection accepts polymer handle that adjusts from 86.4 to 157.5cm (34 to 62 in.)
  • Resistant to water, oil, grease, detergents, sanitizers and solvents
  • Washes and rinses clean with warm water and dries quickly
  • Handle sold separately
Specifications
Product TypeFloor Squeegee
For Use With (Application)Designed for use on uneven or irregular surfaces
MaterialNeoprene Rubber, Polymer Channel
Unit SizePack