Kut Gard™ Cotton-Lined Seamless Knit Hot Mill Glove
Actual product may vary
PIP
Kut Gard™ Cotton-Lined Seamless Knit Hot Mill Glove
Aramid Fiber/Preox blended shell provides resistance against cuts, abrasions, and flames. Gloves protect against the intense heat of an electric arc complying with NFPA 70E standards.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-160-843X-Large
FIS19-160-841Small
Catalog number FIS19-160-843
Price (SGD)
-
Size:
X-Large
  • Aramid Fiber/Preox shell provides cut, abrasion, and flame resistance
  • Preox is a heat and flame resistant fiber that withstands continuous exposure up to 700°F without degradation
  • 100% terry cotton lined for additional comfort and heat protection
  • Seamless construction increases comfort and breathability
  • Ambidextrous design extends wear and reduces replacement costs
  • Extended, open cuff allows for easy on/off and upper arm protection
  • Protects against the intense heat of an electric arc complying with NFPA 70E standards
  • Meets OSHA CFR 1910.269 standards
Specifications
Certifications/ComplianceOSHA CFR 1910.269, NFPA 70E
Accelerator FreeYes
ApplicationsIndustrial
Coating MaterialUncoated
ColorGreen
Cut Resistance RatingANSI A4
HandAmbidextrous
Latex FreeYes
MaterialAramid Fiber, Preox
Product TypeHot Mill Gloves
SizeX-Large
StyleStandard
Temperature (English) Max.700°F
Temperature (Metric) Max370°C
For Use With (Application)Hot Metal Handling, Glass Handling, Foundries, and Forging Operations
Glove Cuff StyleOpen
Liner MaterialCotton
Series43-858
Yarn Weight32 oz.
Unit SizeEach