MICROFLEX™ NeoTouch™ 25-201 Neoprene Gloves, Extended Cuff
Actual product may vary
Ansell
MICROFLEX™ NeoTouch™ 25-201 Neoprene Gloves, Extended Cuff
Neoprene gloves with an extended cuff offers expanded chemical splash protection over the wrist and forearm.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-160-398CLarge
FIS19-160-398BMedium
Catalog number FIS19-160-398C
Price (SGD)
95.00
Pack
Size:
Large
Price (SGD)
95.00
Pack
Manufactured from neoprene. Ansell NeoTouch™ Neoprene Gloves have textured fingertips to provide a secure grip in both dry and wet conditions.
  • Neoprene formulation
  • Textured fingertips provide a secure grip in wet and dry environments
  • Chemical splash resistance to most acids and alcohols
  • Polymer coated for easy donning
  • Extended cuff for expanded protection over the wrist and forearm
Specifications
ANSI Abrasion RatingANSI 0
Certifications/ComplianceASTM D6977, FDA 21 CFR 177-2600
Certifications/Compliance (Europe)EN 374-1:2016, EN 374-5:2016, EN 420:2003 + A1:2009
ChlorinatedYes
ApplicationsLaboratory
AQL1.5
ColorGreen
Exterior FinishPartially Textured
Finger Thickness (English)6.3 mil
Finger Thickness (Metric)0.16 mm
Latex FreeYes
MaterialNeoprene
Palm Thickness (English)5.1 mil
Palm Thickness (Metric)0.13 mm
PowderedPowder Free
Product TypeNeoprene Gloves
SizeLarge
Thickness (English)6.3 mil
Thickness (Metric)0.16 mm
EN 421Yes
EN ISO 374-1Type B
EN ISO 374 - 5Bacteria, Fungi WITH Virus Protection
Glove Cuff StyleExtended
Grip DesignTextured Fingers
LiningUnlined
Model25-201 L 385681
Packaging TypeCase
SeriesMICROFLEX™ NeoTouch™ 25-201
Length (English)11.5 in.
Length (Metric)29 cm
Unit SizePack