HyFlex™ 11-318 Industrial Knit Gloves
Actual product may vary
HyFlex™ 11-318 Industrial Knit Gloves
HyFlex™ 11-318 Industrial Knit Gloves
HyFlex™ 11-318 Industrial Knit Gloves
Ansell
HyFlex™ 11-318 Industrial Knit Gloves
Lightweight industrial knit gloves offer barehand-like comfort and touchscreen compatibility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-152-3068
FIS19-152-3046
FIS19-152-3057
Catalog number FIS19-152-306
Price (SGD)
459.00
Each
Size:
8
Price (SGD)
459.00
Each
  • Industrial safety gloves have an ultralight 18-gauge design, ensuring advanced dexterity and touchscreen compatibility.
  • Silicone-free to prevent potential contaminant transfer to metal prior to painting.
  • Dyneema™ Diamond Technology fiber helps achieve EN ISO B/ANSI A2 cut protection.
  • Spandex™ reinforcement in the palm and knuckles provides a cushioned fit.
  • Oeko-Tex™ certification ensures gloves are free of harmful substances.
  • Recommended for handling parts, assembly, inspection, and maintenance.
Specifications
ANSI Abrasion RatingANSI 2
Certifications/Compliance (Europe)CE, EN 420:2003 + A1:2009; Category II
Coated AreaUncoated
ConstructionKnit
Coating MaterialUncoated
ColorGray
Cut Resistance RatingANSI A2
HandSpecific
Latex FreeYes
MaterialHPPE
Product TypeCut-resistant Gloves
Size8
EN388 StandardEN 388:XX4XB, EN 388:X34X
For Use With (Application)Assembly, Inspection, Automotive, Machinery, Maintenance
GenderUnisex
Glove Cuff ColorGray
Glove Cuff StyleKnit
Liner MaterialDyneema™ Diamond Technology
LiningLined
Model11-318
Puncture RatingANSI/ISEA 105-16 Level 0
SeriesHyFlex™ 11-318
Yarn Gauge18
Length (English)8.3 in.
Length (Metric)21.2 cm
Unit SizeEach