Label Maker Cartridge: Nylon Cloth
Actual product may vary
Fisherbrand
Label Maker Cartridge: Nylon Cloth
Label Maker Cartridge: Nylon Cloth (B-499) wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
Have Questions?
Catalog NumberDimensions (L x W)
FIS19-150-2540.825 x 0.375 in.
Catalog number FIS19-150-254
Price (SGD)
-
Dimensions (L x W):
0.825 x 0.375 in.
Label Maker Cartridge: Nylon Cloth (B-499) wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
  • Matte white polyamide coated nylon cloth material has a permanent acrylic adhesive and is thermal transfer printable
  • Recommended for indoor use
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted
  • All-in-one cartridge includes ribbon and label supply
Specifications
ColorBlack
For Use With (Equipment)BMP51 Label Printer, BMP53 Label Printer
MaterialNylon Cloth
Width (English)0.82 in.
Width (Metric)9.53 mm
Height (English)0.37 in.
Height (Metric)9.5 mm
Thickness (English)0.0065 in.
Thickness (Metric)0.165 mm
Dimensions (L x W)0.825 x 0.375 in.
Adhesive TypePermanent
SterilityNon-sterile
DescriptionBMP41/BMP51/BMP53 Labelmaker Labels
FinishMatte White
IncludesNot Applicable
ShapeRectangle
Tensile Strength80 lb./in
Temperature Range (English)-40°F to +194°F
Temperature Range (Metric)-40°C to +90°C
Certifications/ComplianceUL, CSA, DIN VDE 0472 Part 815, ASTM D
For Use With (Application)Wiremarking and General Labeling Applications
Quantity1 Ea.
Unit SizeEach