Globally Harmonized System (GHS) Flammable Picto Labels
Actual product may vary
Globally Harmonized System (GHS) Flammable Picto Labels
For flammable gases, liquids and solids; pyrophorics; self-reatives; self-heating
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityMaterialHeight (English)Width (English)
FIS19-140-5494/Pk.Vinyl2 in.2 in.
FIS19-140-51339/Pk.Vinyl0.62 in.0.6 in.
FIS19-140-53116/Pk.Vinyl1 in.1 in.
Catalog number FIS19-140-549
Price (SGD)
29.00
Pack
Quantity:
4/Pk.
Material:
Vinyl
Height (English):
2 in.
Width (English):
2 in.
Price (SGD)
29.00
Pack
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards.
  • Legend: Flammable
  • Made of Vinyl or Coated paper
  • Available in three sizes
Specifications
Certifications/ComplianceGHS, OSHA 29 CFR 1910.1200
Adhesive TypePermanent Acrylic
ColorBlack, Red, White
FinishGloss
FormatLabel
MaterialVinyl
Product TypePictogram Label
SterilityNon-sterile
Display TextFlammable
For Use With (Application)Flammable gases, liquids and solids
Quantity4/Pk.
Temperature Range (English)-20°F to 75°F
Temperature Range (Metric)-4°C to 167°C
Dimensions (L x W)2 x 2 in.
Width (English)2 in.
Height (English)2 in.
Height (Metric)5 cm
Unit SizePack