WypAll™ Jumbo Roll Wiper Systems
Actual product may vary
Fisherbrand
WypAll™ Jumbo Roll Wiper Systems
Efficient dispensing system reduces waste
Have Questions?
Catalog NumberProduct Type
FIS19-130-5330Jumbo Roll Dispenser
Catalog number FIS19-130-5330
Price (SGD)
104.00
Case
Product Type:
Jumbo Roll Dispenser
Price (SGD)
104.00
Case
Dispensers are compatible with WypAll X80 Jumbo Roll wipers (sold separately)
Choose from two dispenser styles
  • Wall-Mount Dispenser — Black metal with trashbag attachment; measures 16.75 × 8.75 × 10.75 in. (42.5 × 22.2 × 27.3cm)
    • Self-Standing Dispenser — Portable, high-capacity, black metal dispenser compact enough for use in a variety of locations; measures 16.75 × 33 × 18.5 in. (42.5 × 83.8 × 47cm)
Specifications
Product TypeJumbo Roll Dispenser
ColorBlack
Depth (English)18.5 in.
Depth (Metric)46.99 cm
Width (English)4 in.
Width (Metric)16.8 cm
Height (English)33 in.
Height (Metric)83.82 cm
MaterialMetal
For Use With (Application)Efficient dispensing system reduces waste
For Use With (Equipment)WYPALL™ and KIMTECH™ jumbo roll products
IncludesTrash bag attachment
Unit SizeCase