Antistatic Cleanroom Bags
Actual product may vary
KNF FLEXPAK
Antistatic Cleanroom Bags
Constructed from low-density amine-free polyethylene film
Have Questions?
Catalog NumberDimensions (L x W)Quantity
FIS19-130-214648 x 48 in.1 Ea.
Catalog number FIS19-130-2146
Price (SGD)
45.00
Each
Dimensions (L x W):
48 x 48 in.
Quantity:
1 Ea.
Price (SGD)
45.00
Each
Flat construction.
  • 6mil thick
  • Tough yet flexible
  • Exhibits low water vapor transmission rate and good electrical properties
Specifications
AutoclavableNot Autoclavable
Certifications/ComplianceNASA JPG 5322.1 Level 150. Certificate of conformance accompanies shipment and reflects part number
ColorClear
DisposableSingle-use
MaterialPolyethylene
Quantity1 Ea.
Thickness (English)6 mil
Dimensions (L x W)48 x 48 in.
Length (English)48 in.
Width (English)48 in.
Product TypeCleanroom Bags
Length (Metric)122 cm
Width (Metric)122 cm
Unit SizeEach