WypAll™ X80 Towels
Actual product may vary
WypAll™ X80 Towels
WypAll™ X80 Towels
WypAll™ X80 Towels
WypAll™ X80 Towels
Ansell
WypAll™ X80 Towels
Absorb oil and water faster than rags or shop towels
Have Questions?
Catalog NumberColorDimensions (L x W)FormatPackaging Quantity
FIS19-130-1631Blue16.8 x 12.5 in.BRAG™ box160 Sheets/Pk.
Catalog number FIS19-130-1631
Price (SGD)
144.00
Pack
Color:
Blue
Dimensions (L x W):
16.8 x 12.5 in.
Format:
BRAG™ box
Packaging Quantity:
160 Sheets/Pk.
Price (SGD)
144.00
Pack
Absorb oil and water faster than rags or shop towels
  • Made with patented Hydroknit™ fast-absorbing material and designed to feel like cloth
  • Use for heavy-duty cleaning, wiping parts, collecting metal shavings, prepping surfaces with solvents, and scrubbing rough surfaces
  • Retain strength through harsh use, even when used with solvents
  • Choice of three colors and packaging styles
Specifications
Product TypeHeavy-duty Wiper
Length (English)16.8 in.
Length (Metric)42.67 cm
Width (English)16.8 in.
Width (Metric)31.75 cm
MaterialNon-woven Polyproylene Cellulose
ColorBlue
DisposableSingle-use
Dimensions (L x W)16.8 x 12.5 in.
FormatBRAG™ box
For Use With (Application)General-Purpose
Packaging Quantity160 Sheets/Pk.
Unit SizePack