KleenGuard™ V80 Revolution™ OTG Safety Goggles
Actual product may vary
Ansell
KleenGuard™ V80 Revolution™ OTG Safety Goggles
When you need splash protection, anti-fog lenses and a sleek look rolled into one, trust KleenGuard V80 Revolution OTG Safety Goggles.
Have Questions?
Catalog NumberCertifications/Compliance
FIS19-130-0480ANSI Z87.1-2010
Catalog number FIS19-130-0480
Price (SGD)
45.00
Each
Certifications/Compliance:
ANSI Z87.1-2010
Price (SGD)
45.00
Each
  • Impact-resistant polycarbonate lenses with VisiClear™ antifog, -scratch and -static coating
  • Easy-to-clean material conforms to face
  • Cross Vent™ Technology keeps air flowing in and moisture or chemical splashes out
  • Blue frames with headband
Specifications
Lens TypeVisiClear
Lens CoatingAnti-fog
Certifications/ComplianceANSI Z87.1-2010
Lens TintClear
Product TypeSafety Goggles
Frame ColorBlue
Ventilation DetailsCross vent
MaterialPolycarbonate
Unit SizeEach