Spilfyter™ Formaldehyde Solidifier Spill Kit
Actual product may vary
NPS Corp.
Spilfyter™ Formaldehyde Solidifier Spill Kit
Designed for safe formaldehyde clean up
Have Questions?
Catalog number FIS19-122-877
Price (SGD)
177.00
Each
Price (SGD)
177.00
Each
  • Kit designed for safe clean up of formaldehyde spills
  • Contains formaldehyde solidifier that reduces vapors from formaldehyde spills
  • Size: Small
Specifications
TypeFormaldehyde Spill Kit
For Use With (Application)To provide a safe method of cleaning up and reducing the vapors from formaldehyde leaks and spills
Includes1 formaldehyde solidifier, two 0.9kg shaker bottle, one AQUALOCKIT™ 2 0.9kg shaker bottle, two 12 x 12 in. hazmat pads, one scoop with detachable scraper, one pair nitrile gloves, one pair goggles, one disposal bag and twist tie
SizeSmall
Weight (English)5 lb.
Weight (Metric)2 kg
Unit SizeEach