CleanTeam™ 30-LayerContamination Control Mats
Actual product may vary
PIP
CleanTeam™ 30-LayerContamination Control Mats
Reduce footborne dust and particulates
Have Questions?
Catalog NumberColorDimensions (L x W)
FIS19-122-834Blue66 x 114 cm (26 x 45 in.)
Catalog number FIS19-122-834
Price (SGD)
594.00
Case
Color:
Blue
Dimensions (L x W):
66 x 114 cm (26 x 45 in.)
Price (SGD)
594.00
Case
Reduce footborne dust and particulates
  • Multi-layered construction (0.050mm per layer)
  • Adhesive coating
  • Numbered tabs
  • 30-layer configuration
Specifications
Dimensions (L x W)66 x 114 cm (26 x 45 in.)
DisposableLimited-use
Quantity30 Sheets/Mat
ColorBlue
Length (English)45 in.
Length (Metric)114.3 cm
MaterialPolyethylene
TypeContamination Control Mat
Width (English)45 in.
Width (Metric)114.3 cm
Unit SizeCase