Vinyl Sleeves
Actual product may vary
Ansell
Vinyl Sleeves
Ideal for environments that must be contaminant-free
Have Questions?
Catalog NumberColorThicknessPackaging
FIS19-121-101Clear8 mil12 Pr./Pk.
Catalog number FIS19-121-101
Price (SGD)
81.00
Pack
Color:
Clear
Thickness:
8 mil
Packaging:
12 Pr./Pk.
Price (SGD)
81.00
Pack
  • Sleeves made from vinyl
  • Heat-sealed to prevent contamination problems that come from sewn seams, such as pieces of thread or lint
  • Length: 18 in. (45.7cm)
  • Available in 4mil, 6mil and 8mil thickness in clear or blue
Specifications
MaterialVinyl
ColorClear
Cuff StyleElastic
Thickness8 mil
For Use With (Application)Chemical processing, electronics manufacturing, food processing, food service, forestry, natural gas, oil refining, pharmaceutical
Length (English)18 in.
Length (Metric)45.7 cm
Packaging12 Pr./Pk.
Unit SizePack