Scott™ and Kleenex™ Folded Paper Towels
Actual product may vary
Ansell
Scott™ and Kleenex™ Folded Paper Towels
Offer excellent absorbency and hand-drying performance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)For Use With (Equipment)Packaging Quantity
FIS19-120-4769.4 x 9.2 in.Multi-fold towel dispenser250 Towels/Clip
FIS06-666-32B13.15 x 10.12 in.C-fold Dispenser200 Towels/Clip
Catalog number FIS19-120-476
Price (SGD)
213.00
Case
Dimensions (L x W):
9.4 x 9.2 in.
For Use With (Equipment):
Multi-fold towel dispenser
Packaging Quantity:
250 Towels/Clip
Price (SGD)
213.00
Case
  • Special drying process produces a softer finished towel
  • Reliable quality, softness and absorbency means fewer towels used for better economy
  • Available in C-fold and multifold styles
Specifications
Product TypeMulti-fold Towel
Length (English)9.4 in.
Length (Metric)23.9 cm
Width (English)9.4 in.
Width (Metric)23.36 cm
MaterialPaper
Certifications/ComplianceEPA, FSC™, EcoLogo™
ColorWhite
Dimensions (L x W)9.4 x 9.2 in.
FormatMulti-fold
For Use With (Application)Designed to reduce waste and provides excellent absorbency and hand-drying performance
For Use With (Equipment)Multi-fold towel dispenser
Packaging Quantity250 Towels/Clip
Unit SizeCase