Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Personal Protective Equipment
▸
Gloves
▸
Cleanroom Gloves
▸
MICROFLEX™ CE5 System™ Nitrile Gloves
Actual product may vary
Microflex
MICROFLEX™ CE5 System™ Nitrile Gloves
Exceptional dexterity, tactile sense, fit, feel and comfort.
Have Questions?
Change view
Catalog Number
Size
FIS19-120-2500C
Large
FIS19-120-2500B
Medium
FIS19-120-2500A
Small
3 Options
Catalog number
FIS19-120-2500C
Price (SGD)
988.00
Pack
Add to cart
Size:
Large
Price (SGD)
988.00
Pack
Add to cart
Micro-textured fingertip surface enhances wet or dry grip
Beaded cuff for easier donning
Low surface particulates, ionic extractables and nonvolatile residues
Excellent static decay and surface resistivity properties
Contain no latex, plasticizers or silicone
Min. single wall finger thickness: 4.7 mil
Lengths: 9 and 12 in. (23 and 30.5 cm)
White
Specifications
Color
White
Exterior Finish
Partially Textured
Hand
Ambidextrous
Material
Nitrile
Powdered
Powder Free
Size
Large
Sterility
Non-sterile
Thickness (English)
4.7 mil
Model
CE5-755-L
Length (English)
12 in.
Length (Metric)
30.5 cm