SafeAir™ Detection Badges
Actual product may vary
SafeAir™ Detection Badges
SafeAir™ Detection Badges
SafeAir™ Detection Badges
Morphix Technologies
SafeAir™ Detection Badges
Single-level screening badges provide economical daily exposure monitoring.
Have Questions?
Catalog NumberDetectable AnalytesTemperature Range (English)
FIS19-087519Aromatic Isocyanates60°F to 91°F
Catalog number FIS19-087519
Price (SGD)
965.00
Pack
Detectable Analytes:
Aromatic Isocyanates
Temperature Range (English):
60°F to 91°F
Price (SGD)
965.00
Pack
When the target gas is detected at the threshold level, an immediate color change occurs and an exclamation point is visible within the color triangle on the badge.
  • Provide immediate results
  • Easy to read
  • No laboratory analysis is required
  • May be used for as little as 15 minutes or up to two days; for minimum/maximum recommended sampling times, exposure level, and minimum detectable limit for each analyte, contact your Fisher Safety Customer Service Representative
Specifications
For Use With (Application)Personnel or area monitoring for exposure times ranging from 15 min. to 24 Hours
Detectable AnalytesAromatic Isocyanates
Measurement Range0.4 ppm.Hour
Shelf Life1 Year
Temperature Range (English)60°F to 91°F
Temperature Range (Metric)15.55°C to 32.77°C
Weight (English)0.003 lb.
Weight (Metric)0.001 kg
Product TypeDetection Badge
Unit SizePack