Tyvek™ 400 Frocks, Open Wrists
Actual product may vary
DuPont
Tyvek™ 400 Frocks, Open Wrists
Tyvek frock features a body-length jacket design with front snap closures for easy donning and doffing. Available in individually packaged styles for PPE vending machines.
Have Questions?
Catalog NumberSizePackaging Type
FIS19-087223X-LargeBulk Packaged
Catalog number FIS19-087223
Price (SGD)
559.00
Case
Size:
X-Large
Packaging Type:
Bulk Packaged
Price (SGD)
559.00
Case
Tyvek frock features a body-length jacket design with front snap closures for easy donning and doffing. Available in individually packaged styles for PPE vending machines.
  • Flash spun high density polyethylene nonwoven material
  • Provides ideal balance of protection, durability, and comfort
  • Offers an inherent barrier against particles (down to 1.0 micron in size)
  • Common applications include lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up
  • Frocks provide body-length jacket design with front snap closures for easy donning and doffing
  • Frock extends below hip
  • Laydown collar
Specifications
SizeX-Large
MaterialTyvek
ColorWhite
DisposableLimited-use
Closure TypeSnaps
Cuff StyleOpen
Collar TypeLaydown
StyleBelow Hip-length
Size (English) Chest41.75 to 45.25 in.
Certifications/ComplianceASTM D1777
Packaging TypeBulk Packaged
Length (Metric)99.06 cm
Length (English) Sleeve34.75 in.
Length (Metric) Sleeve88.26 cm
Length (English)39 in.
Product TypeGeneral Purpose Frock
Seam TypeSerged
For Use With (Application)Lead and asbestos abatement and remediation, general maintenance and operations, spray painting, general clean-up
Unit SizeCase