Uvex™ Astro OTG 3001™ Safety Glasses
Actual product may vary
Uvex™ Astro OTG 3001™ Safety Glasses
Over-the-glasses style
Have Questions?
Catalog NumberFrame ColorLens TypeLens TintLens Coating
FIS19-084889BlackUvextreme Anti-fogClearAnti-fog
Catalog number FIS19-084889
Price (SGD)
-
Frame Color:
Black
Lens Type:
Uvextreme Anti-fog
Lens Tint:
Clear
Lens Coating:
Anti-fog
Over-the-glasses style
  • Patented adjustable lens inclination and adjustable temple length offer one-size-fits-all flexibility; Duoflex™ temples have soft comfort cushions for the sensitive mastoid area behind the ear
  • Lightweight, impact-resistant nylon frames feature polycarbonate sideshields and browguard for protection against UV, dust, and flying particles
  • Clear polycarbonate lenses with Ultradura™ hardcoat (UD) that resists scratches and absorbs 99.9% of UV radiation up to 385nm
uvextreme™ Anti-Fog Coating (XTR)
  • Permanently bonded to lens
  • Absorbs 99.9% of UV radiation
  • Anti-scratch and -static
  • Will not wear after repeated cleanings
ultra-dura™ Hard Coat (UD)
  • Permanently bonded to lens
  • Scratch- and chemical-resistant
Specifications
Lens TypeUvextreme Anti-fog
Lens CoatingAnti-fog
Certifications/ComplianceANSI Z87.1-2015, CSA Z94.3, Military V0 Ballistic Test
DescriptionClear XTR
Lens TintClear
Product TypeOver-the-Glasses Safety Spectacles
Frame ColorBlack
Lens MaterialPolycarbonate
Frame MaterialNylon
UV ProtectionUV400
Unit SizeEach