Coventry™ Cleanroom Swabs
Actual product may vary
Chemtronics
Coventry™ Cleanroom Swabs
Have Questions?
Catalog NumberConstructionDiameter (Metric) HandleHandle MaterialTip Material
FIS19-083307Sealed3 mmPolypropyleneKnit Polyester
Catalog number FIS19-083307
Price (SGD)
647.00
Pack
Construction:
Sealed
Diameter (Metric) Handle:
3 mm
Handle Material:
Polypropylene
Tip Material:
Knit Polyester
Price (SGD)
647.00
Pack
Static Dissipative Swabs
  • Employ a plastic alloy that dissipates static in less than two seconds
  • Resist common cleaning solvents including acetone
  • Do not shed particles or rely on humidity to be effective
  • Meet/exceed military and federal test requirements for static dissipation and protection
  • Available with medical grade fine cell polyurethane foam or knitted, cleanroom polyester fabric heads
  • Pack of 500
Sealed Polyester Swabs
  • Resist acids and solvents
  • Cleanroom polyester fabric heads produced by a proprietary thermo-welding process
  • Available with additional Class M1.5 (Class 1) cleaing Aquaprime™ process
Diamond™ Swabs
  • For ultra-critical applications
  • Tubular polyester knit, no-edge, seamless head design
  • Lowest NVR, ionic, and particulate possible
  • Ideal for validating surfaces in biotechnology
  • Pack of 100
Pillow-Tip™ Swabs
  • For applications requiring large solvent retention or a soft, seamless swab
  • Lint-free alternative for cotton swabs
  • Excellent chemical resistance
  • Pack of 25
Wrapped Foam Swabs
  • Fine cell cleanroom medical grade foam
  • Pointed spiral design for precision cleaning and fluid application
  • Available with wood or tough nylon handles
  • Pack of 500
Sealed Foam Swabs
  • Economical general purpose swabs
  • Highly absorbent open-cell structure
  • Fiberless construction and free of adhesives and binders
  • Polypropylene handles
  • Pack of 500
Specifications
ConstructionSealed
Diameter (English) Handle0.12 in.
Length (English)5.8 in.
For Use With (Application)Can be used to validate biogenetic/pharmaceutical surfaces, clean disk drive components, clean sensitive surfaces in optics, clean aerospace hardware
Product TypeSwab
Diameter (Metric) Handle3 mm
Length (Metric)147 mm
MaterialKnit Polyester
Thickness (Metric)4.8 mm
Length (English) Tip0.75 in.
Length (Metric) Tip18.9 mm
Width (Metric) Tip5 mm
Width (English) Tip0.4 in.
Handle MaterialPolypropylene
Tip MaterialKnit Polyester
Unit SizePack