EcoPolyBlend™ Spill Tray
Actual product may vary
Justrite
EcoPolyBlend™ Spill Tray
Lightweight and low-profile design for small-scale spill control
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)Depth (English)Width (English)
FIS19-075-35323 gal.34 in.37.75 in.
FIS19-075-35012 gal.16 in.46 in.
Catalog number FIS19-075-353
Price (SGD)
432.00
Each
Capacity (English):
23 gal.
Depth (English):
34 in.
Width (English):
37.75 in.
Price (SGD)
432.00
Each
  • Eco-friendly: made of 100% recycled polyethylene
  • Will not rust or corrode
  • Raised rib bottom to add rigidity
  • Rated for service temperatures between -34 to +49°C (-30° to +120°F)
Specifications
ColorBlack
Capacity (English)23 gal.
Capacity (Metric)87 L
MaterialPolyethylene
TypeSpill Tray
For Use With (Application)Perfect for pouring and filling applications
ResistantChemical
Width (English)37.75 in.
Width (Metric)95.88 cm
Height (English)5.5 in.
Height (Metric)13.97 cm
Depth (English)34 in.
Depth (Metric)86.36 cm
Unit SizeEach