▸
Facility Safety and Maintenance
▸
Spill Control and Containment
▸
Secondary Containment
▸
EcoPolyBlend™ Spill Tray
Actual product may vary
Justrite
EcoPolyBlend™ Spill Tray
Lightweight and low-profile design for small-scale spill control
Have Questions?
Change view
Catalog Number
Capacity (English)
Depth (English)
Width (English)
FIS19-075-350
12 gal.
16 in.
46 in.
FIS19-075-353
23 gal.
34 in.
37.75 in.
2 Options
Catalog number
FIS19-075-350
Price (SGD)
282.00
Each
Add to cart
Capacity (English):
12 gal.
Depth (English):
16 in.
Width (English):
46 in.
Price (SGD)
282.00
Each
Add to cart
Eco-friendly: made of 100% recycled polyethylene
Will not rust or corrode
Raised rib bottom to add rigidity
Rated for service temperatures between -34 to +49°C (-30° to +120°F)
Specifications
Color
Black
Capacity (English)
12 gal.
Capacity (Metric)
45 L
Material
Polyethylene
Type
Spill Tray
For Use With (Application)
Perfect for pouring and filling applications
Resistant
Chemical
Width (English)
46 in.
Width (Metric)
116.8 cm
Height (English)
5.5 in.
Height (Metric)
13.97 cm
Depth (English)
16 in.
Depth (Metric)
40.64 cm