Cotton-Tipped Applicators
Actual product may vary
Puritan
Cotton-Tipped Applicators
Mini cotton swab features a miniature tapered cotton tip and sturdy wooden handle used for general cleaning. Available sterile or non-sterile.
Have Questions?
Catalog NumberLength (English)Sterility
FIS19-062-7103 in.Non-sterile
Catalog number FIS19-062-710
Price (SGD)
723.00
Case
Length (English):
3 in.
Sterility:
Non-sterile
Price (SGD)
723.00
Case
General cleaning swab;Ideal for hard-to-reach areas; Made in USA
Specifications
Certifications/ComplianceISO 9001:2008, ISO 13485:2003, FDA GMP's
Diameter (English) Handle0.083 in.
Length (English)3 in.
For Use With (Application)General cleaning swab, for hard-to-reach areas
Product TypeApplicator
Diameter (Metric) Handle2.11 mm
Length (Metric)7.62 cm
Length (English) Handle2.9 in.
Length (Metric) Handle7.3 cm
Length (English) Tip0.5 in.
Length (Metric) Tip12.7 mm
Diameter (Metric) Tip3.05 mm
Diameter (English) Tip0.12 in.
SterilityNon-sterile
Handle MaterialWood
Tip StyleTapered Mini
Tip MaterialCotton
Unit SizeCase