100% Cotton Lisle Inspector's Gloves, Long Length
Actual product may vary
MCR Safety
100% Cotton Lisle Inspector's Gloves, Long Length
Thin, 100% cotton lisle inspection gloves are reversible and unhemmed. 8614C gloves feature an extended, 14 in. length for additional coverage.
Have Questions?
Catalog number FIS19-058833
Price (SGD)
22.00
Pack
Price (SGD)
22.00
Pack
  • Reversible
  • Unhemmed
  • 100% Cotton
  • Straight thumb
  • Long, 14 in. length
  • Comfortable and light weight
  • Breathable fabric helps hands cool
  • Wear alone, or use as glove liners
Specifications
ConstructionString Knit
Cuff MaterialCotton
ColorWhite
GradeGeneral Protection
HandAmbidextrous
MaterialCotton
Product TypeInspection Gloves
SizeLarge
StyleReversible
Glove Cuff ColorWhite
Glove Cuff StyleStraight
Thumb StyleStraight
Length (English)14 in.
Length (English) Cuff11 in.
Length (Metric)35.6 cm
Length (Metric) Cuff28 cm
Unit SizePack