Posi M3 Sleeves
Actual product may vary
West Chester
Posi M3 Sleeves
Five layers of protection providing a high level of breathability and protection against dry particulates and water based light liquid sprays and splashes. Good for use in industrial manufacturing, paper industry, food processing and any heavy dust environment.
Have Questions?
Catalog NumberSize
FIS19-055-158Universal
Catalog number FIS19-055-158
Price (SGD)
291.00
Case
Size:
Universal
Price (SGD)
291.00
Case
Five layers of protection For use in industrial manufacturing, paper industry, food processing and any heavy dust environment
Specifications
Product TypeGeneral Purpose Sleeve
SizeUniversal
MaterialPolyethylene, Spunbond Polypropylene
ColorWhite
SterilityNon-sterile
DisposableSingle-use
Closure TypeElastic Ends
Seam TypeSerged
For Use With (Application)Industrial manufacturing, paper industry, food processing
Length (English)18 in.
Length (Metric)45.7 cm
Unit SizeCase