Neoprene Sleeve VSW-HS 59-050
Actual product may vary
Ansell
Neoprene Sleeve VSW-HS 59-050
Neoprene sleeve features elastic on both ends and is ideal for a wide range of chemical applications.
Have Questions?
Catalog NumberCuff Style
FIS19-053-730Elastic
Catalog number FIS19-053-730
Price (SGD)
24.00
Each
Cuff Style:
Elastic
Price (SGD)
24.00
Each
  • Double-insulating thermal liner: Resistance to both hot and cold
  • High-grade neoprene coating: Advanced mechanical and chemical resistance
  • Flexible, grip-enhancing design: Maneuverability and confident handling
N/A
Specifications
SizeStandard
MaterialNeoprene
ColorYellow
SterilityNon-sterile
DisposableReusable
Closure TypeElastic Ends
Cuff StyleElastic
FinishSealed
Latex FreeNo
Model59-050
Thickness16.9 mil
For Use With (Application)Chemical Processing, Electronics Manufacturing, Food Processing, Forestry, Oil Refining, Pharmaceutical
Length (English)18 in.
Length (Metric)45.7 cm
Packaging12 Pairs/Bag, 8 Bags/Cs.
Unit SizeEach