CSM Glove Box Gloves, 8 in. Port
Actual product may vary
Honeywell
CSM Glove Box Gloves, 8 in. Port
CSM glove box gloves feature mechanical properties to protect against abrasion, tears, and ozone and oxidizing chemicals.
Have Questions?
Catalog NumberSizeHandThickness (Metric)
FIS19-050-20710.5Ambidextrous0.76 mm
Catalog number FIS19-050-207
Price (SGD)
1,926.00
Pair
Size:
10.5
Hand:
Ambidextrous
Thickness (Metric):
0.76 mm
Price (SGD)
1,926.00
Pair
CSM glove box gloves feature mechanical properties to protect against abrasion, tears, and ozone and oxidizing chemicals.
  • CSM gloves for use in glove boxes
  • Abrasion resistant
  • Resistant to ozone or oxidizing chemicals
  • Naturally soft for worker comfort
  • White surface for enhanced visibility
  • Hand specific or ambidextrous
Specifications
Certifications/Compliance (Europe)EN 388, EN 374
ColorWhite
HandAmbidextrous
Latex FreeYes
MaterialCSM
Size10.5
SterilityNon-sterile
Thickness (English)30 mil
Thickness (Metric)0.76 mm
Grip DesignSmooth
Model8Y3032A/10H
Port Size (English)8 in.
Port Size (Metric)20.3 cm
Length (English)32 in.
Length (Metric)81.3 cm
Diameter (English) Cuff8 in.
Diameter (Metric) Cuff20.3 cm
Unit SizePair