Kimtech™ A8 Lab Coats, Traditional Collar
Actual product may vary
Kimtech™ A8 Lab Coats, Traditional Collar
Kimtech™ A8 Lab Coats, Traditional Collar
Ansell
Kimtech™ A8 Lab Coats, Traditional Collar
Non-woven SMS lab coat provides comfort and protection from light fluid contact.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-0430612X-Large
KLC10122Large
KLC10120Small
KLC10121Medium
KLC10123X-Large
Catalog number FIS19-043061
Price (SGD)
603.00
Case
Size:
2X-Large
Price (SGD)
603.00
Case
  • Protective three-layer white SMS fabric resistant to blood strike-through for various levels of exposure
  • Feature traditional collar, knit cuffs, four snaps, two reinforced pockets, and size/name label
OSHA requires the employer to evaluate the task and type of exposure anticipated and select the appropriate clothing that prevents contamination of an employee's skin or clothing by blood or other potentially infectious materials
Specifications
Product TypeGeneral Purpose Lab Coat
Size2X-Large
GenderUnisex
MaterialSMS
ColorWhite
Closure TypeSnaps
Cuff StyleKnit
DisposableLimited-use
SterilityNon-sterile
ResistanceFluids
No. of Pockets2
Collar TypeTraditional
Model10124
Size (English) Chest54 to 60 in.
Thickness3-ply
Construction3-ply SMS
Unit SizeCase