Scott™ Essential Hard Roll Towels
Actual product may vary
Ansell
Scott™ Essential Hard Roll Towels
High-footage roll towel that provides reliable quality that lasts. Highly absorbent, so you’ll use less and waste less. Compatible with most universal hard roll towel dispensers Elemental chlorine-free (ECF) bleaching. Meets EPA standards, FSC™ and EcoLogo™ certified.
Have Questions?
Catalog NumberColor
FIS19-043-220White
Catalog number FIS19-043-220
Price (SGD)
509.00
Case
Color:
White
Price (SGD)
509.00
Case
  • 800 feet per roll, 12 rolls per case for a total of 9,600 feet
  • High capacity: reduce maintenance time, run-outs and complaints with long-lasting bulk paper towels (as compared to a folded towel system)
  • Compatible with many Kimberly-Clark Professional hard roll paper towel dispensers, along with many universal hard roll paper towel dispensers designed to hold 1.5 inch cores
  • Highly absorbent commercial paper towels let you and your bathroom guests use less and waste less
  • Paper towels come in space-saving cases for easy storage and handling at your business
Specifications
Product TypeHard Roll Towel
Length (English)800 ft.
Length (Metric)24,384 cm
Width (English)800 ft.
Width (Metric)20.32 cm
MaterialPaper
AbsorbancyHighly absorbent
Certifications/ComplianceEPA, FSC™, EcoLogo™
ColorWhite
Dimensions (L x W)800 ft. x 8 in. (243.84 m x 20.32 cm)
For Use With (Application)Provides reliable quality that lasts
For Use With (Equipment)Most universal hard roll towel dispensers
Ply1-ply
Unit SizeCase