AlphaTec™ 56-402 Heavy Duty Neoprene Apron
Actual product may vary
Ansell
AlphaTec™ 56-402 Heavy Duty Neoprene Apron
Black heavy duty neoprene apron provides chemical, oil, grease, puncture, and abrasion resistance.
Have Questions?
Catalog NumberNo. per Pack
FIS19-042-2661 Ea.
Catalog number FIS19-042-266
Price (SGD)
702.00
Dozen
No. per Pack:
1 Ea.
Price (SGD)
702.00
Dozen
  • 16-17 mil sewn-edge neoprene: Resists chemicals, oils, punctures, and abrasion
  • Nylon reinforcement: For a superb balance of comfort and personal protection
  • Sealed grommets and two-piece 48-inch poly ties: Adjustable, consistent fit
N/A
Specifications
TypeApron
SizeLarge
MaterialNeoprene
ColorBlack
DisposableSingle-use
Closure TypeTies
SterilityNon-sterile
ConstructionNeoprene, Sewn Edge Finish
FinishSewn-edge
Latex FreeYes
No. of PocketsNone
ResistanceChemical Splash, Oils, Grease, Punctures, Abrasion
Thickness16 to 17 mil
For Use With (Application)Food Service, Maintenance, Transferring Liquids
Dimensions (L x W)45 x 35 in.
Length (English)45 in.
Length (Metric)114.3 cm
Width (English)35 in.
Width (Metric)88.9 cm
Weight (English)12.5 oz.
No. per Case12/Cs.
No. per Pack1 Ea.
Seam TypeSewn
Unit SizeDozen