UltraSoft Diamond-Plate Mat
Actual product may vary
Wearwell
UltraSoft Diamond-Plate Mat
Antifatigue mat designed for ultimate comfort and performance
Have Questions?
Catalog NumberDimensions (L x W)Color
FIS19-042-2283 x 2 ft.Black/Yellow
Catalog number FIS19-042-228
Price (SGD)
242.00
Each
Dimensions (L x W):
3 x 2 ft.
Color:
Black/Yellow
Price (SGD)
242.00
Each
Antifatigue mat designed for ultimate comfort and performance
  • For heavy use areas, mats resist abrasion and most chemical spills
  • Double thick sponge base made from 0.94 in. (2.4cm) immediate recovery Nitricell™ sponge
  • Specially formulated blend of nitrile rubber and PVC
  • Slip resistant, diamond embossed vinyl surface resists chemicals, abrasion and fire
  • All edges are beveled for safety and convenience
  • Recycled: Item contains recycled content as described in the item detail
Specifications
Dimensions (L x W)3 x 2 ft.
ColorBlack/Yellow
Length (Metric)91.4 cm
MaterialDouble-thick Nitricell Sponge
Width (Metric)61 cm
Unit SizeEach