VisionAid™ SafetyClean Lens-Cleaning Towelettes
Actual product may vary
VisionAid™ SafetyClean Lens-Cleaning Towelettes
VisionAid™ SafetyClean Lens-Cleaning Towelettes
Raidians
VisionAid™ SafetyClean Lens-Cleaning Towelettes
Antifog, antistatic premoistened towelettes
Have Questions?
Catalog NumberProduct Type
FIS19-041-637Cleaning Wipe
Catalog number FIS19-041-637
Price (SGD)
20.00
Pack
Product Type:
Cleaning Wipe
Price (SGD)
20.00
Pack
Prevent fogging and reduce static with these individually packaged, premoistened towelettes.
  • Each foil-wrapped packet contains one 5 × 8 in. (12.7 × 20.3cm) Lensclean™ tissue premoistened with Lensclean antifog, antistatic cleaning solution
  • Towelettes are silicone-free and safe for use on both glass and plastic lenses
  • Sold in dispenser packs of 100 towelettes
Specifications
DescriptionLens Cleaning Towelettes
Product TypeCleaning Wipe
For Use With (Equipment)Glass and plastic lenses
For Use With (Application)Prevent fogging and reduce static
No. per Pack100 towelettes
Unit SizePack