R95 Particulate Respirator, 8240
R95 Particulate Respirator, 8240
Actual product may vary
Fisherbrand
R95 Particulate Respirator, 8240
Economical R95 disposable particulate respirator features an adjustable nose clip and cushioning foam to provide a custom and secure seal against certain oil and non oil based particles.
Have Questions?
Catalog number FIS19-039789
Price (SGD)
-
Economical R95 disposable particulate respirator features an adjustable nose clip and cushioning foam to provide a custom and secure seal against certain oil and non oil based particles.
  • NIOSH approved R95 rating
  • Adjustable nose clip
  • Nose foam
  • Braided headbands
Specifications
Aerosol TypeR - Somewhat Resistant to Oil
Assigned Protection Factor (APF)10
Certifications (NIOSH)NIOSH 42 CFR 84, R95, approval number: TC-84A-1350
Certifications (OSHA)OSHA 1910.134
ColorWhite
Design TypeCup/Molded
DisposableSingle-use
Filter TypeR95
MaterialPolypropylene, Polyester
Product TypeGeneral Purpose Particulate Respirator
SizeStandard
With Exhalation ValveNo
Filter Efficiency95%
For Use With (Application)Foundry Operations, Grinding, Petrochemical Manufacturing, Processing of Minerals, Welding
Latex FreeYes
Material HeadstrapPolyisoprene, Polyester
Model8240
Nose Piece MaterialAluminum
Nosepiece TypeAdjustable Nose Clip
Quantity120/Cs.
Shelf Life5 years from date of manufacture
SterilityNon-sterile
With FacesealNo
Strap Attachment TypeBraided Headbands
FDA Cleared Surgical MaskNo
Unit SizeCS