Polypropylene Floor-Mounted Acid Cabinet
Actual product may vary
Polypropylene Floor-Mounted Acid Cabinet
Keep acids safe and secure. The SciMat™ Polypropylene Acid Cabinet is floor-mounted, providing extra worker protection by keeping acids further from the face.
Have Questions?
Catalog NumberHeight (English) ExteriorHeight (Metric) ExteriorNo. of Shelves
FIS19-039-78160 in.152.4 cm3
Catalog number FIS19-039-781
Price (SGD)
-
Height (English) Exterior:
60 in.
Height (Metric) Exterior:
152.4 cm
No. of Shelves:
3
White polypropylene construction will not rust or corrode; Interior seams are welded liquid-tight, meeting OSHA's secondary containment standards; Available in two heights.
Constructed of .38 in. thick (0.95cm) white polypropylene with .5 in. thick doorsfor years of service in even the most corrosive environments. Even the hinges are made of polypropylene for rust-free, corrosion-free durability. All interior seams are welded for the liquid-tight seal needed to meet OSHA's secondary containment standards. A 2 in. (5.1cm) trough contains internal spills, while shelves are lipped to provide further liquid containment, to prevent roll-off of bottles, and to add structural strength.
Specifications
Certifications/ComplianceOSHA
Depth (Metric) Exterior47 cm
Door StyleManual-close
Height (Metric) Exterior152.4 cm
IncludesLock and keys, 2 in. trough contains internal spills, liquid tight all-welded interior to contain accidental spills
Load Bearing Capacity66 gal.
No. of Doors2
No. of Shelves3
Width (Metric) Exterior121.92 cm
ColorWhite
Depth (English) Exterior18.5 in.
Height (English) Exterior60 in.
MaterialPolypropylene
Product TypeAcids and Alkalis Cabinet (Floor Model)
Width (English) Exterior48 in.
Unit SizeEach