4-in-1 Absorbent Mat Rolls in Dispenser Box
Actual product may vary
4-in-1 Absorbent Mat Rolls in Dispenser Box
4-in-1 Absorbent Mat Rolls in Dispenser Box
New Pig
4-in-1 Absorbent Mat Rolls in Dispenser Box
This versatile mat roll can also be used as a pad, sock or wiper
Have Questions?
Catalog NumberCapacity (English)
FIS19-039-1709.2 gal.
Catalog number FIS19-039-170
Price (SGD)
307.00
Each
Capacity (English):
9.2 gal.
Price (SGD)
307.00
Each
  • Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job
  • Dispenser box makes mat easy to access, carry and store while protecting contents from dirt and moisture
  • Rolls are perforated every 25.4cm (10 in.); easy to tear to size
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Highly absorbent, fine-fiber construction won't leave liquids or fiber residue behind
Specifications
ColorGray
Capacity (English)9.2 gal.
Capacity (Metric)34.8 L
MaterialPolypropylene
TypePIG™ 4-in-1™ Absorbent Mat Roll
For Use With (Application)Absorbs oils, coolants, solvents and water
Dimensions (L x W)80 ft. x 16 in. (24.4 m x 40.6 cm)
Length (English)80 ft.
Length (Metric)24.4 m
Width (English)16 in.
Width (Metric)40.6 cm
Weight (English)10 lb.
Weight (Metric)4.53 kg
Unit SizeEach