Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Furniture
▸
Shelving
▸
Shelving Posts and Accessories
▸
Super Adjustable Super Erecta™ Wire Shelving Accessory, 8 in. Shelf Dividers
Actual product may vary
Metro
Super Adjustable Super Erecta™ Wire Shelving Accessory, 8 in. Shelf Dividers
Tall dividers (8 in.) snap onto wire shelves to partition shelf space for storage optimization.
Have Questions?
Catalog Number
Width (English)
FIS19-037480A
18 in.
Catalog number
FIS19-037480A
Price (SGD)
89.00
Each
Add to cart
Width (English):
18 in.
Price (SGD)
89.00
Each
Add to cart
Dividers can also attach to the top or bottom of shelves.
Available in Chrome or Type 304 Stainless Steel.
Other sizes available.
Specifications
For Use With (Equipment)
Metro™ Super Erecta™ shelving
Height (English)
8 in.
Height (Metric)
20.3 cm
Width (English)
18 in.
Width (Metric)
45.7 cm
Product Type
Shelf Divider for Wire Shelves
Material
Chrome