▸
Facility Safety and Maintenance
▸
Cleaning Supplies and Equipment
▸
Cleaners and Disinfectants
▸
General Cleaning Products
▸
Cleanroom Squeegee with Swivel
Actual product may vary
Vileda Professional
Cleanroom Squeegee with Swivel
Swivels 360 degrees allowing for full range of motion
Have Questions?
Catalog Number
Length (English)
FIS19-032-1041
20 in.
Catalog number
FIS19-032-1041
Price (SGD)
186.00
Each
Add to cart
Length (English):
20 in.
Price (SGD)
186.00
Each
Add to cart
Use on floors and walls after applying disinfectant
Squeegee spills to the drain in controlled environments
Can be autoclaved and exposed to gamma or EtO sterilization
Use with FHP Extendable Handle #119966
Specifications
Product Type
Swivel Squeegee
For Use With (Application)
Cleanroom applications
Length (English)
20 in.