KC100 Kimguard™ Sterilization Wraps
KC100 Kimguard™ Sterilization Wraps
Actual product may vary
KC100 Kimguard™ Sterilization Wraps
Soft, drapable wrap withstands tough handling
Have Questions?
Catalog NumberDimensions (L x W)
FIS19-02988112 x 12 in. (30.4 x 30.4 cm)
Catalog number FIS19-029881
Price (SGD)
-
Dimensions (L x W):
12 x 12 in. (30.4 x 30.4 cm)
Soft, drapable wrap withstands tough handling
First of six strength levels in the Kimguard line is often used for light linen packs and gowns, towel packs, light basins and other lighter wrapping needs.
  • Soft, strong and tear-resistant
  • Resists event-related contamination
  • Tough three-layer, low-lint SMS construction resists penetration by particles and liquids
  • Naturally latex-free, nonsterile, and steam and EtO compatible
  • Color: Light blue
Specifications
ColorLight Blue
For Use With (Application)Used to package linen towels, packs, gowns, light basin sets and single instruments for sterilization
MaterialSMS Fabric
SterilityNon-sterile
Latex FreeYes
Resistant ToLint
Sterilization MethodSteam and EtO
Dimensions (L x W)12 x 12 in. (30.4 x 30.4 cm)
Length (English)12 in.
Length (Metric)30.4 cm
Width (English)12 in.
Width (Metric)30.4 cm
Unit SizeCS