Phlebotomy Sharps Collectors
Actual product may vary
BD
Phlebotomy Sharps Collectors
Built-in needle removal ports designed to prevent contact with sharps during disposal
Have Questions?
Catalog NumberCapacity (English)Closure ColorDimensions (L x W x H)
FIS19-0278761.5 qt.Natural4.75 x 4 x 9 in.
Catalog number FIS19-027876
Price (SGD)
-
Capacity (English):
1.5 qt.
Closure Color:
Natural
Dimensions (L x W x H):
4.75 x 4 x 9 in.
  • Disposable polypropylene
  • Clear tops for clear view of fill level
  • Temporary and final closures for safety and security
  • Cat. No. 19-027-876 features dual access
Specifications
Capacity (Metric)1.4 L
Closure TypeLockable
Closure ColorNatural
Capacity (English)1.5 qt.
ColorRed
Dimensions (L x W x H)4.75 x 4 x 9 in.
DisposableSingle-use
Height (Metric)22.3 cm
Length (Metric)12 cm
VentYes
Width (Metric)10.2 cm
AutoclavableAutoclavable
For Use With (Application)Phlebotomy
Width (English)4 in.
Length (English)4.75 in.
Height (English)9 in.