▸
Facility Safety and Maintenance
▸
Hand Tools and Power Tools
▸
Scissors
▸
Grafco™ Operating Scissors
Actual product may vary
Graham-Field
Grafco™ Operating Scissors
Stainless steel operating scissors
Have Questions?
Catalog Number
Tip Style
FIS19-027491
Sharp/Sharp
Catalog number
FIS19-027491
Price (SGD)
12.00
Each
Add to cart
Tip Style:
Sharp/Sharp
Price (SGD)
12.00
Each
Add to cart
Straight
Length: 14cm (5.5 in.)
Stainless-steel
Specifications
Blade Style
Straight
Length (English)
5.5 in.
Length (Metric)
14 cm
For Use With (Application)
Surgical, Precision
Tip Style
Sharp/Sharp