Multi-Flex™ CC100 Non-Sterile Latex Cleanroom Gloves
Multi-Flex™ CC100 Non-Sterile Latex Cleanroom Gloves
Actual product may vary
Cardinal Health
Multi-Flex™ CC100 Non-Sterile Latex Cleanroom Gloves
Powder free, 12 in. latex gloves meet all relevant ASTM standards and are recommended for Class 100/ISO 5 or higher cleanrooms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-0251616.5
FIS19-065-3357
FIS19-0251658
Catalog number FIS19-025161
Price (SGD)
775.00
Case
Add to cart
Size:
6.5
Price (SGD)
775.00
Case
Add to cart
  • Powder free, natural rubber latex
  • Recommended for ISO 5/Class 100 or higher critical environments
  • Clean-processed and chlorinated
  • Interlocking, beaded cuff design helps reduce roll-down
  • Exclusive hand molds with independent thumb design allow for natural fit and reduces hand fatigue
Specifications
Certifications/ComplianceASTM D3577, IEST-RP-CC005
Cleanroom ClassClass 100 (ISO 5)
AQL1
ColorNatural
Exterior FinishNo Texture
Finger Thickness (English)8 mil
Finger Thickness (Metric)0.20 mm
HandSpecific
Latex FreeNo
MaterialLatex
Palm Thickness (English)7.7 mil
Palm Thickness (Metric)0.19 mm
PowderedPowder Free
Size6.5
SterilityNon-sterile
Thickness (English)8 mil
Thickness (Metric)0.20 mm
Manufacturing/QMS Audit StandardsISO 13485
Model2Y1602
SeriesMulti-Flex™ CC100
Tensile Strength34 MPa
Length (English)12 in.
Length (Metric)30.5 cm
Unit SizeCase