Kimtech™ A8 Certified Lab Coats with Knit Cuffs and Collar
Kimtech™ A8 Certified Lab Coats with Knit Cuffs and Collar
Actual product may vary
Kimtech™ A8 Certified Lab Coats with Knit Cuffs and Collar
Breathable SMS lab coats protect from light fluid contact and feature soft knit cuffs and 3 pockets. Choose from white or blue colors.
Have Questions?
Catalog NumberColorSizeSize (English) Chest
FIS19-022846BlueMedium36 to 40 in.
Catalog number FIS19-022846
Price (SGD)
814.00
CS
Add to cart
Color:
Blue
Size:
Medium
Size (English) Chest:
36 to 40 in.
Price (SGD)
814.00
CS
Add to cart
  • Lab coats with knit cuffs and collar
  • Comfort and protection from light fluid contact
  • 3-layer SMS nonwoven fabric
  • Soft knit cuffs and collar
  • Mid-calf length
  • Three reinforced pockets and five snaps
  • Neck loop for convenient hanging
Specifications
Product TypeGeneral Purpose Lab Coat
SizeMedium
GenderUnisex
MaterialSMS
ColorBlue
Closure TypeSnaps
Cuff StyleKnit
DisposableLimited-use
SterilityNon-sterile
StyleMid-calf Length
No. of Pockets3
Collar TypeKnit
Model10031
Size (English) Chest36 to 40 in.
Thickness3-ply
IncludesHanging Neck Loop
Construction3-ply SMS
Unit SizeCS