Spilfyter™ Mercury Spill Kit
Actual product may vary
NPS Corp.
Spilfyter™ Mercury Spill Kit
Designed to handle all stages of mercury spill cleanup
Have Questions?
Catalog number FIS19-021910
Price (SGD)
444.00
Each
Price (SGD)
444.00
Each
Designed to handle all stages of mercury spill cleanup
Kit contains everything needed to quickly and safely clean up mercury spills up to 0.75 oz. (25mL) and prevent toxic vapor contamination.
  • Mercury indicator powder identifies mercury in area of suspected contamination
  • After undisturbed period, powder changes color to indicate contact with mercury metal or vapor
  • Mercsorb™ powder converts mercury into amalgam for safe, convenient cleanup of mercury on work surfaces, floors and hard-to-reach areas
Specifications
Capacity (English)0.75 oz.
Capacity (Metric)25 mL
TypeMercury Spill Kit
For Use With (Application)Chemical spills
Includes270 g amalgamation powder, 250 g indicator powder, 1 vapor suppressor shaker bottle, 1 aspirator bottle, 1 waste collection bottle, mixing tub with spatula, 1 chemical sponge, 1 disposal bag and twist tie, 1 pair nitrile gloves, 1 pair splash goggles
Weight (English)5 lb.
Weight (Metric)2 kg
Unit SizeEach