Tychem™ 2000 Sleeved Apron
Actual product may vary
Tychem™ 2000 Sleeved Apron
Tychem™ 2000 Sleeved Apron
DuPont
Tychem™ 2000 Sleeved Apron
QC Sleeved Aprons offer added chemical protection from numerous light splash situations for a variety of industries.
Have Questions?
Catalog NumberSize
FIS19-019337Universal
Catalog number FIS19-019337
Price (SGD)
25.00
Each
Size:
Universal
Price (SGD)
25.00
Each
  • Knee-length apron/gown design with attached sleeves
  • Available in multiple sizes
  • Provides enhanced frontal protection
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Open back for wearer comfort
  • Bound seams around neck opening with snap closure
  • Ties at waist
  • Elastic opening for tighter fit at wrist
  • 44 in. long for neck-to-knee coverage
  • Manufactured under specifications that do not contain natural rubber latex
Specifications
SizeUniversal
MaterialTychem
ColorYellow
Closure TypeWaist Ties
Cuff StyleElastic
Thickness10 mil
Length (English)52 in.
Length (Metric)132.08 cm
Seam TypeBound
Unit SizeEach