Jomac™ Kelklave Autoclave Gloves
Actual product may vary
Wells Lamont
Jomac™ Kelklave Autoclave Gloves
Jomac™ Kelklave Autoclave Gloves featuring loop-out terrycloth construction are comfortable and flexible, while also providing protection from temperatures up to 250°F.
Have Questions?
Catalog NumberLength (English) Cuff
FIS19-01358611 in.
Catalog number FIS19-013586
Price (SGD)
32.00
Pair
Length (English) Cuff:
11 in.
Price (SGD)
32.00
Pair
Made from high-loop terry cloth, a natural insulator
  • Loop-out terry cloth is flexible and comfortable
  • Gauntlet cuff extends burn protection up the arm
  • Heat resistance up to 250°F
  • Ideal for autoclaves and metal handling
Specifications
ColorWhite
MaterialTerrycloth
Product TypeAutoclave Gloves
SizeLarge
StyleClute
Temperature (English) Max.250°F
Temperature (Metric) Max120°C
For Use With (Application)Metal Handling
Glove Cuff StyleGauntlet
Series422
Length (English) Cuff11 in.
Length (Metric) Cuff28 cm
Unit SizePair